Benzenesulfonyl chloride ≥99.0% (by GC, titration analysis)
Supplier: TCI America
|
Danger
|
Synonyms:
Benzene sulphochloride
B0036-25G
B0036-500G
TCB0036-025GEA
17.12
USD
TCB0036-025G
TCB0036-500G
Benzenesulfonyl chloride ≥99.0% (by GC, titration analysis)
Benzenesulphonyl chloride
CAS Number: 98-09-9
MDL Number: MFCD00007426
Molecular Formula: C6H5ClO2S
Molecular Weight: 176.61
Purity/Analysis Method: >99.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 252
Flash Point (°C): 135
Freezing point (°C): 15
Specific Gravity (20/20): 1.38
Formula:
C₆H₅ClO₂S MW: 176.62 g/mol Boiling Pt: 252 °C Melting Pt: 15 °C Density: 1.38 (20 °C) Flash Pt: 135 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007426 CAS Number: 98-09-9 EINECS: 202-636-6 UN: 2225 ADR: 8,III Merck Index: 13,01072 |
Specification Test Results
Purity/Analysis Method | >99.0% (GC,T) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...