Phenyl phosphate ≥99.0% (by titrimetric analysis)
Supplier: TCI America
CAS Number: 701-64-4
MDL Number: MFCD00058970
Molecular Formula: C6H7O4P
Molecular Weight: 174.09
Purity/Analysis Method: >99.0% (T)
Form: Crystal
Melting point (°C): 100
Other Product Names / Synonyms: Phosphoric Acid Phenyl Ester, Dihydrogen Phenyl Phosphate
Formula:
C₆H₇O₄P Melting Pt: 100 °C |
MDL Number:
MFCD00058970 CAS Number: 701-64-4 |
|
Specification Test Results
Purity/Analysis Method | >99.0% (T) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...