2-Amino-3-hydroxybenzoic acid ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: 3-Hydroxyanthranilic acid

A0316-1G A0316-5G
TCA0316-001GEA 136 USD
TCA0316-001G TCA0316-005G
2-Amino-3-hydroxybenzoic acid ≥98.0% (by HPLC, titration analysis)
2-Amino-3-hydroxybenzoic acid

CAS Number: 548-93-6
MDL Number: MFCD00007700
Molecular Formula: C7H7NO3
Molecular Weight: 153.14
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 240


Other Product Names / Synonyms: 2-Amino-3-hydroxybenzoic Acid

Formula: C₇H₇NO₃
MW: 153.14 g/mol
Boiling Pt: 357 °C (1013 hPa)
Melting Pt: 240 °C
MDL Number: MFCD00007700
CAS Number: 548-93-6
EINECS: 208-962-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=85dfe100343ac4d679ce18177e654a6deea6376e&vid=85dfe100343ac4d679ce18177e654a6deea6376e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR