2,3-Naphthalenedialdehyde (NDA) ≥99.0%
Supplier: TCI America
Synonyms:
Naphthalene-2,3-dicarbaldehyde, Naphthalene-2,3-dicarboxaldehyde, NDA
A5594-100MG
A5594-1G
TCA5594-100MGEA
138
USD
TCA5594-100MG
TCA5594-1G
2,3-Naphthalenedialdehyde (NDA) ≥99.0%
2,3-Naphthalenedialdehyde (NDA)
[Fluorimetric Reagent for Primary Amines]
CAS Number: 7149-49-7
MDL Number: MFCD00016618
Molecular Formula: C12H8O2
Molecular Weight: 184.19
Purity/Analysis Method:
Formula:
C₁₂H₈O₂ MW: 184.19 g/mol Melting Pt: 133 °C |
MDL Number:
MFCD00016618 CAS Number: 7149-49-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...