Xylometazoline hydrochloride ≥98.0% (by HPLC, total nitrogen)
Supplier: TCI America
Synonyms:
2-(4-(tert-Butyl)-2,6-dimethylbenzyl)-4,5-dihydro-1H-imidazole hydrochloride, 2-(4-tert-Butyl-2,6-dimethylbenzyl)-2-imidazoline Hydrochloride
X0063-5G
X0063-1G
TCX0063-5GEA
113
USD
TCX0063-5G
TCX0063-1G
Xylometazoline hydrochloride ≥98.0% (by HPLC, total nitrogen)
Xylometazoline hydrochloride
CAS Number: 1218-35-5
MDL Number: MFCD00238707
Molecular Formula: C16H24N2
Molecular Weight: 280.84
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Color: White
Product Categories: Bioscience, Other Pharmaceutical Ingredients
Other Product Names / Synonyms: 2-(4-tert-Butyl-2,6-dimethylbenzyl)-2-imidazoline Hydrochloride
Formula:
C₁₆H₂₄N₂ MW: 280.84 g/mol |
MDL Number:
MFCD00238707 CAS Number: 1218-35-5 |
|
Specification Test Results
Purity/Analysis Method | >98.0% (HPLC,N) |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...