p-Xylylene dithiocyanate ≥98.0%
Supplier: TCI America
Synonyms:
alpha,alpha'-Dithiocyanato-p-xylene
X0060-25G
X0060-5G
TCX0060-25GEA
387
USD
TCX0060-25G
TCX0060-5G
p-Xylylene dithiocyanate ≥98.0%
p-Xylylene dithiocyanate
CAS Number: 1014-99-9
Molecular Formula: C10H8N2S2
Molecular Weight: 220.31
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: White
Melting point (°C): 137
Other Product Names / Synonyms: alpha,alpha'-Dithiocyanato-p-xylene
Formula:
C₁₀H₈N₂S₂ MW: 220.31 g/mol Melting Pt: 137 °C |
CAS Number:
1014-99-9 |
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...