m-Xylylene diisocyanate ≥98.0%

Supplier: TCI America

Synonyms: 1,3-Bis(isocyanatomethyl)benzene, Isocyanic Acid 1,3-Phenylenedimethylene Ester, m-Xylene Diisocyanate, M-xylylenediisocyanate

X0022-500G X0022-25G X0022-100G
TCX0022-500GEA 326.45 USD
TCX0022-500G TCX0022-025G TCX0022-100G
m-Xylylene diisocyanate ≥98.0%
m-Xylylene diisocyanate

CAS Number: 3634-83-1
MDL Number: MFCD00019917
Molecular Formula: C10H8N2O2
Molecular Weight: 188.19
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 126
Melting point (°C): -7
Flash Point (°C): 174
Specific Gravity (20/20): 1.20
Storage Temperature: 0-10°C


Product Categories: Materials Chemistry, Monomers, Polyimide Raw Materials
Other Product Names / Synonyms: 1,3-Bis(isocyanatomethyl)benzene, Isocyanic Acid 1,3-Phenylenedimethylene Ester, m-Xylene Diisocyanate

Formula: C₁₀H₈N₂O₂
MW: 188.19 g/mol
Boiling Pt: 126 °C
Melting Pt: –7 °C
Density: 1.20 (20 °C)
Flash Pt: 174 °C
MDL Number: MFCD00019917
CAS Number: 3634-83-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=78c564535da4754a5db9a1b354075857e9dad5bd&vid=78c564535da4754a5db9a1b354075857e9dad5bd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR