Vinylphosphonic acid ≥95.0% (by titrimetric analysis)
Supplier: TCI America
CAS Number: 1746-03-8
MDL Number: MFCD00043866
Molecular Formula: C2H5O3P
Molecular Weight: 108.03
Purity/Analysis Method: >95.0% (T)
Form: Clear Liquid
Color: Very Pale Yellow
Boiling point (°C): 98
Flash Point (°C): 110
Specific Gravity (20/20): 1.40
Formula:
C₂H₅O₃P Boiling Pt: 98 °C Density: 1.40 (20 °C) Flash Pt: 110 °C |
MDL Number:
MFCD00043866 CAS Number: 1746-03-8 UN: 3265 ADR: 8,III |
|
Specification Test Results
Purity/Analysis Method | >95.0% (T) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...