5-Vinylbicyclo[2.2.1]hept-2-ene ≥98.0% stabilized

Supplier: TCI America

Synonyms: 5-Vinyl-2-norbornene

V0062-500ML V0062-25ML
TCV0062-500MLEA 95 USD
TCV0062-500ML TCV0062-025ML
5-Vinylbicyclo[2.2.1]hept-2-ene ≥98.0% stabilized
5-Vinylbicyclo[2.2.1]hept-2-ene

(stabilized with BHT)
CAS Number: 3048-64-4
MDL Number: MFCD00080691
Molecular Formula: C9H12
Molecular Weight: 120.20
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 141
Flash Point (°C): 24
Specific Gravity (20/20): 0.89


Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: 5-Vinyl-2-norbornene

Formula: C₉H₁₂
MW: 120.20 g/mol
Boiling Pt: 141 °C
Density: 0.89 (20 °C)
Flash Pt: 24 °C
MDL Number: MFCD00080691
CAS Number: 3048-64-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=26e7d0f110e3e96598a7f91c488db015f503bf90&vid=26e7d0f110e3e96598a7f91c488db015f503bf90&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR