Vincamine ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: Vincamine base, Methyl (41S,12S,13aS)-13a-ethyl-12-hydroxy-2,3,41,5,6,12,13,13a-octahydro-1H-indolo[3,2,1-de]pyrido[3,2,1-ij][1,5]naphthyridine-12-carboxylate

V0061-1G V0061-5G
TCV0061-001GEA 85.44 USD
TCV0061-001G TCV0061-005G
Vincamine ≥98.0% (by titrimetric analysis)
Vincamine

CAS Number: 1617-90-9
MDL Number: MFCD00078054
Molecular Formula: C21H26N2O3
Molecular Weight: 354.45
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: White
Melting point (°C): 232
Specific rotation [a]20/D: 43 deg (C=1, Pyridine)

Formula: C₂₁H₂₆N₂O₃
MW: 354.45 g/mol
Melting Pt: 232 °C
MDL Number: MFCD00078054
CAS Number: 1617-90-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=04c4023fe5c4095343feff64067c5386dd130989&vid=04c4023fe5c4095343feff64067c5386dd130989&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR