Methylstyrene (mixture of isomers) ≥98.0% stabilized
Supplier: TCI America
V0028-25ML
V0028-500ML
TCV0028-025MLEA
30.1
USD
TCV0028-025ML
TCV0028-500ML
Methylstyrene (mixture of isomers) ≥98.0% stabilized
Methylstyrene (mixture of isomers)
(stabilized with TBC)
CAS Number: 25013-15-4
MDL Number: MFCD00168066
Molecular Formula: C9H10
Molecular Weight: 118.18
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 173
Melting point (°C): -77
Flash Point (°C): 53
Specific Gravity (20/20): 0.90
Storage Temperature: 0-10°C
Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: Methylstyrene Monomer
Formula:
C₉H₁₀ MW: 118.18 g/mol Boiling Pt: 173 °C Melting Pt: -77 °C Density: 0.90 (20 °C) Flash Pt: 53 °C |
MDL Number:
MFCD00168066 CAS Number: 25013-15-4 EINECS: 246-562-2 UN: 2618 ADR: 3,III |
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...