Undecafluorohexanoic acid ≥98.0% (by titrimetric analysis)

Supplier: TCI America

U0067-5G U0067-25G
TCU0067-005GEA 84.64 USD
TCU0067-005G TCU0067-25G
Undecafluorohexanoic acid ≥98.0% (by titrimetric analysis)
Undecafluorohexanoic acid

CAS Number: 307-24-4
MDL Number: MFCD00198040
Molecular Formula: C6HF11O2
Molecular Weight: 314.05
Purity/Analysis Method: >98.0% (T)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 157
Specific Gravity (20/20): 1.76


Product Categories: Synthetic Organic Chemistry, Fluorous Compounds, Materials Chemistry, Fluorosurfactants
Other Product Names / Synonyms: Perfluorohexanoic Acid

Formula: C₆HF₁₁O₂
Boiling Pt: 157 °C
Density: 1.76 (20 °C)
MDL Number: MFCD00198040
CAS Number: 307-24-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7c4bd588352884b7bb37e4ba637fe4a436cd528f&vid=7c4bd588352884b7bb37e4ba637fe4a436cd528f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR