(±)-3-Undecanol ≥94.0%

Supplier: TCI America

Synonyms: (±)-3-Hydroxy undecane, 3-Undecanol

U0028-5ML U0028-25ML
TCU0028-005MLEA 127 USD
TCU0028-005ML TCU0028-025ML
(±)-3-Undecanol ≥94.0%
(±)-3-Undecanol

CAS Number: 6929-08-4
MDL Number: MFCD00046730
Molecular Formula: C11H24O
Molecular Weight: 172.31
Purity/Analysis Method: >94.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 132
Flash Point (°C): 38
Specific Gravity (20/20): 0.83

Formula: C₁₁H₂₄O
MW: 172.31 g/mol
Boiling Pt: 132 °C
Density: 0.83 (20 °C)
Flash Pt: 38 °C
MDL Number: MFCD00046730
CAS Number: 6929-08-4
EINECS: 230-054-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e1859437cbbb8659828420bded8bcb632500294f&vid=e1859437cbbb8659828420bded8bcb632500294f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >94.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR