(+)-Usnic acid ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: (9bR)-2,6-Diacetyl-7,9-dihydroxy-8,9b-dimethyldibenzo[b,d]furan-1,3(2H,9bH)-dione

U0023-1G U0023-25G
TCU0023-001GEA 24 USD
TCU0023-001G TCU0023-025G
(+)-Usnic acid ≥98.0% (by HPLC, titration analysis)
(+)-Usnic acid

CAS Number: 7562-61-0
MDL Number: MFCD00016878
Molecular Formula: C18H16O7
Molecular Weight: 344.32
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Color: Slightly Pale Yellow
Melting point (°C): 200
Specific rotation [a]20/D: 495 deg (C=0.7, CHCl3)

Formula: C₁₈H₁₆O₇
MW: 344.32 g/mol
Melting Pt: 200 °C
MDL Number: MFCD00016878
CAS Number: 7562-61-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=63cac09f4deb196ae00c8b8c800c7d3ec161d152&vid=63cac09f4deb196ae00c8b8c800c7d3ec161d152&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR