Undec-10-enoyl chloride ≥98.0% (by titrimetric analysis)
Supplier: TCI America
|
Danger
|
CAS Number: 38460-95-6
MDL Number: MFCD00000772
Molecular Formula: C11H19ClO
Molecular Weight: 202.72
Purity/Analysis Method: >98.0% (T)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 93
Flash Point (°C): 93
Specific Gravity (20/20): 0.95
Formula:
C₁₁H₁₉ClO Boiling Pt: 93 °C Density: 0.95 (20 °C) Flash Pt: 93 °C |
MDL Number:
MFCD00000772 CAS Number: 38460-95-6 EINECS: 253-951-0 UN: 3265 ADR: 8,II |
|
Specification Test Results
Purity/Analysis Method | >98.0% (T) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...