Tributylmethylammonium bis(trifluoromethanesulfonyl)imide ≥98.0%

Supplier: TCI America

Synonyms: N,N-Dibutyl-N-methylbutan-1-aminium bis((trifluoromethyl)sulphonyl)amide

T2679-5G T2679-25G
TCT2679-5GEA 143 USD
TCT2679-5G TCT2679-25G
Tributylmethylammonium bis(trifluoromethanesulfonyl)imide ≥98.0%
Tributylmethylammonium bis(trifluoromethanesulfonyl)imide

CAS Number: 405514-94-5
Molecular Formula: C15H30F6N2O4S2
Molecular Weight: 480.53
Purity/Analysis Method: >98.0% (T)
Form: Clear Liquid
Color: Colorless
Specific Gravity (20/20): 1.27

Formula: C₁₅H₃₀F₆N₂O₄S₂
MW: 480.53 g/mol
Density: 1.27 (20 °C)
MDL Number: MFCD12761447
CAS Number: 405514-94-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=855d8748f0b4623b7e38fe393bccb17f83b54e60&vid=855d8748f0b4623b7e38fe393bccb17f83b54e60&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR