L-Threonine benzyl ester hydrochloride ≥98.0% (by HPLC, total nitrogen)
Supplier: TCI America
Synonyms:
(2S,3R)-Benzyl 2-amino-3-hydroxybutanoate hydrochloride, H-Thr-OBzl.HCl
T2677-1G
T2677-5G
TCT2677-1GEA
37
USD
TCT2677-1G
TCT2677-5G
L-Threonine benzyl ester hydrochloride ≥98.0% (by HPLC, total nitrogen)
L-Threonine benzyl ester hydrochloride
CAS Number: 33645-24-8
Molecular Formula: C11H15NO3
Molecular Weight: 245.70
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Color: White
Melting point (°C): 130
Specific rotation [a]20/D: -11 deg (C=1, EtOH)
Storage Temperature: 0-10°C
Product Categories: Bioscience, Amino Acids (C-Protected)
Other Product Names / Synonyms: H-Thr-OBzl.HCl
Formula:
C₁₁H₁₅NO₃ MW: 245.70 g/mol Melting Pt: 130 °C |
MDL Number:
MFCD00038977 CAS Number: 33645-24-8 |
Specification Test Results
Purity/Analysis Method | >98.0% (HPLC,N) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...