Tetrabutylammonium p-nitrophenoxide ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

T2669-25G T2669-5G
TCT2669-25GEA 104 USD
TCT2669-25G TCT2669-5G
Tetrabutylammonium p-nitrophenoxide ≥98.0% (by HPLC, titration analysis)
Tetrabutylammonium p-nitrophenoxide

CAS Number: 3002-48-0 Molecular Formula: C22H40N2O3 Molecular Weight: 380.57 Purity/Analysis Method: 98.0% (HPLC,T) Form: Crystal Color: Very Pale Yellow Melting point (°C): 145

Formula: C₂₂H₄₀N₂O₃
Melting Pt: 145 °C
CAS Number: 3002-48-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=27ce664bcf3cb3b22a6964cb51b636c830b1517c&vid=27ce664bcf3cb3b22a6964cb51b636c830b1517c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR