Tetrakis(acetonitrile)copper(I) tetrafluoroborate ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: Copper(I) tetra(acetonitrile) tetrafluoroborate, Tetrakis(acetonitrile)copper(I) tetrafluoroborate

T2666-5G T2666-1G
TCT2666-5GEA 64 USD
TCT2666-5G TCT2666-1G
Tetrakis(acetonitrile)copper(I) tetrafluoroborate ≥98.0% (by titrimetric analysis)
Tetrakis(acetonitrile)copper(I) tetrafluoroborate

CAS Number: 15418-29-8 MDL Number: MFCD09265110 Molecular Formula: C8H12BCuF4N4 Molecular Weight: 314.56 Purity/Analysis Method: 98.0% (T) Form: Crystal Color: White Melting point (°C): 164

Formula: C₈H₁₂BCuF₄N₄
MW: 314.56 g/mol
Melting Pt: 164 °C
MDL Number: MFCD09265110
CAS Number: 15418-29-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b1cee47f8b7f21b92ce4b03c78b73e16c9493a94&vid=b1cee47f8b7f21b92ce4b03c78b73e16c9493a94&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR