Triphenylbismuth difluoride ≥95.0%

Supplier: TCI America

T2657-1G T2657-5G
TCT2657-1GEA 182 USD
TCT2657-1G TCT2657-5G
Triphenylbismuth difluoride ≥95.0%
Triphenylbismuth difluoride

CAS Number: 2023-48-5
Molecular Formula: C18H15BiF2
Molecular Weight: 478.30
Purity/Analysis Method: >95.0% (T)
Form: Crystal
Color: White
Melting point (°C): 159
Storage Temperature: 0-10°C


Product Categories: Synthetic Organic Chemistry, Hypervalent Bismuth Compounds
Other Product Names / Synonyms: Difluorotriphenylbismuth

Formula: C₁₈H₁₅BiF₂
Melting Pt: 159 °C
CAS Number: 2023-48-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=777f3f530c19641618ceb3beb304474850f0960a&vid=777f3f530c19641618ceb3beb304474850f0960a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >95.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR