N-Tetradecyl-N,N-dimethyl-3-ammonio-1-propanesulphonate (Sulfobetaine-14, SB-14) ≥98.0% (by total nitrogen basis) for biochemical research
Supplier: TCI America
Synonyms:
3-(N,N-Dimethylmyristylammonio)propanesulphonate, Tetradecyldimethyl(3-sulphopropyl)ammonium Hydroxide Inner Salt
[for Biochemical Research]
CAS Number: 14933-09-6
MDL Number: MFCD00036910
Molecular Formula: C19H41NO3S
Molecular Weight: 363.60
Purity/Analysis Method: >98.0% (N)
Form: Crystal
Color: White
Melting point (°C): 245
Flash Point (°C): 110
Product Categories: Bioscience, Amphoteric Surfactants (for Biochemistry)
Other Product Names / Synonyms: 3-[Dimethyl(tetradecyl)ammonio]propane-1-sulfonate, Myristyldimethyl(3-sulfopropyl)ammonium Hydroxide Inner Salt, Myristyl Sulfobetaine, 3-[Dimethyl(myristyl)ammonio]propane-1-sulfonate
Formula:
C₁₉H₄₁NO₃S MW: 363.5988 g/mol Melting Pt: 245 °C Flash Pt: 110 °C |
MDL Number:
MFCD00036910 CAS Number: 14933-09-6 |
Specification Test Results
Purity/Analysis Method | >98.0% (N) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...