2-Thiophenemethanethiol ≥95.0%

Supplier: TCI America

Synonyms: 2-(Mercaptomethyl)thiophene, Thenyl Mercaptan

T2613-5G T2613-1G
TCT2613-5GEA 291 USD
TCT2613-5G TCT2613-1G
2-Thiophenemethanethiol ≥95.0%
2-Thiophenemethanethiol

CAS Number: 6258-63-5
MDL Number: MFCD00107135
Molecular Formula: C5H6S2
Molecular Weight: 130.22
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 86
Specific Gravity (20/20): 1.20


Other Product Names / Synonyms: Thenyl Mercaptan, 2-(Mercaptomethyl)thiophene

Formula: C₅H₆S₂
MW: 130.22 g/mol
Boiling Pt: 86 °C
Density: 1.20 (20 °C)
MDL Number: MFCD00107135
CAS Number: 6258-63-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5c5c8722c674b906b9753d73fc2e7111d5cc9669&vid=5c5c8722c674b906b9753d73fc2e7111d5cc9669&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR