Tetrakis(2,4-pentanedionato)titanium(IV) ∼63% in isopropanol (IPA)
Supplier: TCI America
Synonyms:
Titanium(IV) Acetylacetonate, Acetylacetone Titanium(IV) Salt
T2610-500G
T2610-25G
TCT2610-500GEA
196
USD
TCT2610-500G
TCT2610-25G
Tetrakis(2,4-pentanedionato)titanium(IV) ∼63% in isopropanol (IPA)
Tetrakis(2,4-pentanedionato)titanium(IV)
CAS Number: 17501-79-0
MDL Number: MFCD00045004
Molecular Formula: C20H28O8Ti
Molecular Weight: 444.30
Form: Clear Liquid
Color: Yellow
Other Product Names / Synonyms: Acetylacetone Titanium(IV) Salt, Titanium(IV) Acetylacetonate
Formula:
C₂₀H₂₈O₈Ti MW: 444.30 g/mol |
MDL Number:
MFCD00045004 CAS Number: 17501-79-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...