2,3,3-Trimethyl-3H-benzo[g]indole ≥98.0%

Supplier: TCI America

Synonyms: 2,3,3-Trimethylnaphtho[1,2-b]pyrrole

T2601-25G T2601-5G
TCT2601-25GEA 420 USD
TCT2601-25G TCT2601-5G
2,3,3-Trimethyl-3H-benzo[g]indole ≥98.0%
2,3,3-Trimethyl-3H-benzo[g]indole

CAS Number: 74470-85-2
MDL Number: MFCD00723378
Molecular Formula: C15H15N
Molecular Weight: 209.29
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Color: Pale Yellow
Boiling point (°C): 127
Melting point (°C): 78


Other Product Names / Synonyms: 2,3,3-Trimethylnaphtho[1,2-b]pyrrole

Formula: C₁₅H₁₅N
MW: 209.29 g/mol
Boiling Pt: 127 °C
Melting Pt: 78 °C
MDL Number: MFCD00723378
CAS Number: 74470-85-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0b6669b7f1bdb8f7160a582d08b24fbddafc179c&vid=0b6669b7f1bdb8f7160a582d08b24fbddafc179c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR