N-[2-(Trimethylsilyl)ethoxycarbonyloxy]succinimide ≥98.0% (by total nitrogen basis)
Supplier: TCI America
Synonyms:
2,5-Dioxopyrrolidin-1-yl (2-(trimethylsilyl)ethyl) carbonate, Teoc-OSu
T2591-1G
T2591-5G
TCT2591-1GEA
43
USD
TCT2591-1G
TCT2591-5G
N-[2-(Trimethylsilyl)ethoxycarbonyloxy]succinimide ≥98.0% (by total nitrogen basis)
N-[2-(Trimethylsilyl)ethoxycarbonyloxy]succinimide
CAS Number: 78269-85-9
MDL Number: MFCD02683467
Molecular Formula: C10H17NO5Si
Molecular Weight: 259.33
Purity/Analysis Method: >98.0% (N)
Form: Crystal
Color: White
Melting point (°C): 100
Storage Temperature: 0-10°C
Product Categories: Synthetic Organic Chemistry, Protection & Derivatization Reagents (for Synthesis)
Other Product Names / Synonyms: Teoc-OSu
Formula:
C₁₀H₁₇NO₅Si MW: 259.33 g/mol Melting Pt: 100 °C |
MDL Number:
MFCD02683467 CAS Number: 78269-85-9 |
Specification Test Results
Purity/Analysis Method | >98.0% (N) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...