2,4,6-Trimethyl-2,4,6-trivinylcyclotrisilazane ≥97.0% (by titrimetric analysis)

Supplier: TCI America

T2556-5G T2556-25G
TCT2556-5GEA 68 USD
TCT2556-5G TCT2556-25G
2,4,6-Trimethyl-2,4,6-trivinylcyclotrisilazane ≥97.0% (by titrimetric analysis)
2,4,6-Trimethyl-2,4,6-trivinylcyclotrisilazane

CAS Number: 5505-72-6 MDL Number: MFCD00054934 Molecular Formula: C9H21N3Si3 Molecular Weight: 255.54 Purity/Analysis Method: 97.0% (T) Form: Clear Liquid Color: Colorless Boiling point (°C): 65 Flash Point (°C): 85 Specific Gravity (20/20): 0.96

Formula: C₉H₂₁N₃Si₃
Boiling Pt: 65 °C
Density: 0.96 (20 °C)
Flash Pt: 85 °C
MDL Number: MFCD00054934
CAS Number: 5505-72-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=addd3bbcdaac8d8c60aa5c9c636c9fe2d42b7497&vid=addd3bbcdaac8d8c60aa5c9c636c9fe2d42b7497&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR