Torsemide ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: Torasemide, N-(Isopropylcarbamoyl)-4-(m-tolylamino)pyridine-3-sulphonamide

T2538-5G T2538-1G
TCT2538-5GEA 408 USD
TCT2538-5G TCT2538-1G
Torsemide ≥98.0% (by HPLC, titration analysis)
Torsemide

CAS Number: 56211-40-6
MDL Number: MFCD00866166
Molecular Formula: C16H20N4O3S
Molecular Weight: 348.42
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Color: White
Melting point (°C): 161


Product Categories: Bioscience, Other Pharmaceutical Ingredients
Other Product Names / Synonyms: N-(Isopropylcarbamoyl)-4-(m-tolylamino)pyridine-3-sulfonamide, Torasemide

Formula: C₁₆H₂₀N₄O₃S
MW: 348.42 g/mol
Melting Pt: 161 °C
MDL Number: MFCD00866166
CAS Number: 56211-40-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0d6de0f83ffabc4ac471c90bb9a0c8375498eb0b&vid=0d6de0f83ffabc4ac471c90bb9a0c8375498eb0b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR