Tizanidine hydrochloride ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 5-Chloro-N-(4,5-dihydro-1H-imidazol-2-yl)benzo[c][1,2,5]thiadiazol-4-amine hydrochloride, Tizanidine HCl, 5-Chloro-4-[(4,5-dihydro-1H-imidazol-2-yl)amino]-2,1,3-benzothiadiazole Hydrochloride

T2527-5G T2527-25G
TCT2527-5GEA 67 USD
TCT2527-5G TCT2527-25G
Tizanidine hydrochloride ≥98.0% (by titrimetric analysis)
Tizanidine hydrochloride

CAS Number: 64461-82-1
MDL Number: MFCD00798231
Molecular Formula: C9H8ClN5S
Molecular Weight: 290.17
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: White
Melting point (°C): 280


Product Categories: Bioscience, Neurotransmitter Agent Ingredients
Other Product Names / Synonyms: 5-Chloro-4-[(4,5-dihydro-1H-imidazol-2-yl)amino]-2,1,3-benzothiadiazole Hydrochloride

Formula: C₉H₈ClN₅S
MW: 290.17 g/mol
Melting Pt: 280 °C
MDL Number: MFCD00798231
CAS Number: 64461-82-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e7aa66c97cc07e7c2d4e3dee33dc35e4a9f62e96&vid=e7aa66c97cc07e7c2d4e3dee33dc35e4a9f62e96&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR