Dansyl hydrazide ≥97.0% for synthesis
Supplier: TCI America
Synonyms:
Dansyl hydrazine, 5-Dimethylaminonaphthalene-1-sulphonyl hydrazine, 5-(Dimethylamino)naphthalene-1-sulphonohydrazide
A5552-5G
A5552-1G
TCA5552-005GEA
379
USD
TCA5552-005G
TCA5552-001G
Dansyl hydrazide ≥97.0% for synthesis
Dansyl hydrazide
[for HPLC Labeling]
CAS Number: 33008-06-9
MDL Number: MFCD00003986
Molecular Formula: C12H15N3O2S
Molecular Weight: 265.33
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Product Categories: Analytical Science, Fluorescence Detection (HPLC Labeling Reagents)
Other Product Names / Synonyms: 5-Dimethylaminonaphthalene-1-sulfonyl Hydrazine
Formula:
C₁₂H₁₅N₃O₂S |
MDL Number:
MFCD00003986 CAS Number: 33008-06-9 EINECS: 251-337-7 |
|
Specification Test Results
Purity/Analysis Method | >97.0% (T) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...