L-Tyrosine benzyl ester-p-toluenesulfonate ≥98.0%

Supplier: TCI America

Synonyms: L-Tyrosine benzyl ester p-toluenesulphonate salt, H-Tyr-OBzl·TosOH, H-Tyr-OBzl.Tos-OH

T2443-25G T2443-5G
TCT2443-25GEA 60 USD
TCT2443-25G TCT2443-5G
L-Tyrosine benzyl ester-p-toluenesulfonate ≥98.0%
L-Tyrosine benzyl ester-p-toluenesulfonate

CAS Number: 53587-11-4
MDL Number: MFCD00038959
Molecular Formula: C16H17NO3
Molecular Weight: 443.51
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Color: White
Melting point (°C): 177
Specific rotation [a]20/D: -13 deg (C=3, MeOH)


Product Categories: Bioscience, Amino Acids (C-Protected)
Other Product Names / Synonyms: H-Tyr-OBzl.Tos-OH

Formula: C₁₆H₁₇NO₃
Melting Pt: 177 °C
MDL Number: MFCD00038959
CAS Number: 53587-11-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3f2d85d02db4d646c65685ebb33847c69042ed03&vid=3f2d85d02db4d646c65685ebb33847c69042ed03&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR