4,5,9,10-Tetrahydropyrene ≥95.0%

Supplier: TCI America

T2264-1G T2264-100MG
TCT2264-1GEA 496.9 USD
TCT2264-1G TCT2264-100MG
4,5,9,10-Tetrahydropyrene ≥95.0%
4,5,9,10-Tetrahydropyrene

CAS Number: 781-17-9
MDL Number: MFCD00267195
Molecular Formula: C16H14
Molecular Weight: 206.29
Purity/Analysis Method: >95.0% (GC)
Form: Crystal
Color: White

Formula: C₁₆H₁₄
MW: 206.29 g/mol
MDL Number: MFCD00267195
CAS Number: 781-17-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f10776ec5cd52c0c1dae3fbd9e2be35c4e54659d&vid=f10776ec5cd52c0c1dae3fbd9e2be35c4e54659d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR