p,p,p-Tritolylphosphate 98,0
Supplier: TCI America
Synonyms:
p,p,p-TCP, p,p,p-Tricresyl phosphate, Tris(4-methylphenyl) phosphate, Tri-p-tolyl phosphate, Tritolyl Phosphate
CAS Number: 78-32-0
Molecular Formula: C21H21O4P
Molecular Weight: 368.37
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: White
Boiling point (°C): 430
Melting point (°C): 77
Flash Point (°C): 210
Product Categories: Materials Chemistry, Secondary Battery Research Reagents, Resin Additives
Other Product Names / Synonyms: Phosphoric Acid Tri-p-cresyl Ester, Phosphoric Acid Tri-p-tolyl Ester, Tri-p-tolyl Phosphate
Formula:
C₂₁H₂₁O₄P MW: 368.37 Boiling Pt: 439 Melting Pt: 77 Density: 1.247 Flash Pt: 210 °C |
CAS Number:
78-32-0 UN: 3082 ADR: 9 III |
|
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...