4-(Trifluoromethyl)salicylic acid ≥98.0% (by GC, titration analysis)
Supplier: TCI America
Synonyms:
2-Hydroxy-4-(trifluoromethyl)benzoic acid
T2064-25G
T2064-5G
TCT2064-25GEA
47
USD
TCT2064-25G
TCT2064-5G
4-(Trifluoromethyl)salicylic acid ≥98.0% (by GC, titration analysis)
4-(Trifluoromethyl)salicylic acid
CAS Number: 328-90-5
Molecular Formula: C8H5F3O3
Molecular Weight: 206.12
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Color: White
Melting point (°C): 178
Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 2-Hydroxy-4-(trifluoromethyl)benzoic Acid
Formula:
C₈H₅F₃O₃ MW: 206.1187 g/mol Melting Pt: 178 °C |
MDL Number:
MFCD01529657 CAS Number: 328-90-5 |
Specification Test Results
Purity/Analysis Method | >98.0% (GC,T) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...