3,5,6-Trifluoro-2-hydroxypyridine ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 3,5,6-Trifluoro-2-pyridinol, 3,5,6-Trifluoro-2-pyridone

T2061-1G T2061-5G
TCT2061-1GEA 36 USD
TCT2061-1G TCT2061-5G
3,5,6-Trifluoro-2-hydroxypyridine ≥98.0% (by GC, titration analysis)
3,5,6-Trifluoro-2-hydroxypyridine

CAS Number: 75777-49-0
MDL Number: MFCD04114299
Molecular Formula: C5H2F3NO
Molecular Weight: 149.07
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Color: White
Melting point (°C): 100


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 3,5,6-Trifluoro-2-pyridinol, 3,5,6-Trifluoro-2-pyridone

Formula: C₅H₂F₃NO
MW: 149.07 g/mol
Melting Pt: 100 °C
MDL Number: MFCD04114299
CAS Number: 75777-49-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3663651d178cb0d61138d607bf74f3796d37d20a&vid=3663651d178cb0d61138d607bf74f3796d37d20a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR