Bis(carboxymethyl)trithiocarbonate ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: Trithiocarbonic Acid Bis(carboxymethyl) Ester

T2036-25G T2036-5G
TCT2036-25GEA 593 USD
TCT2036-25G TCT2036-5G
Bis(carboxymethyl)trithiocarbonate ≥98.0% (by titrimetric analysis)
Bis(carboxymethyl)trithiocarbonate

CAS Number: 6326-83-6
MDL Number: MFCD00004357
Molecular Formula: C5H6O4S3
Molecular Weight: 226.28
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: Pale Yellow
Melting point (°C): 173


Other Product Names / Synonyms: Trithiocarbonic Acid Bis(carboxymethyl) Ester

Formula: C₅H₆O₄S₃
MW: 226.28 g/mol
Melting Pt: 173 °C
MDL Number: MFCD00004357
CAS Number: 6326-83-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=43fb22d113590e47c87180c90dae36ea0a6908f3&vid=43fb22d113590e47c87180c90dae36ea0a6908f3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR