2,2',4-Trimethoxybenzophenone ≥98.0%
Supplier: TCI America
T2028-25G
T2028-5G
TCT2028-25GEA
692.6
USD
TCT2028-25G
TCT2028-5G
2,2',4-Trimethoxybenzophenone ≥98.0%
2,2',4-Trimethoxybenzophenone
CAS Number: 33077-87-1
MDL Number: MFCD01098829
Molecular Formula: C16H16O4
Molecular Weight: 272.30
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: White
Boiling point (°C): 200
Melting point (°C): 56
Formula:
C₁₆H₁₆O₄ Boiling Pt: 200 °C Melting Pt: 56 °C |
MDL Number:
MFCD01098829 CAS Number: 33077-87-1 |
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...