Tetraethylene glycol monobenzyl ether ≥95.0% (by GC)

Supplier: TCI America

Synonyms: BnO-PEG4-OH, Tetraethylene Glycol Monobenzyl Ether, 2-[2-[2-[2-(Benzyloxy)ethoxy]ethoxy]ethoxy]ethanol

T1997-5G T1997-25G
TCT1997-5GEA 201 USD
TCT1997-5G TCT1997-25G
Tetraethylene glycol monobenzyl ether ≥95.0% (by GC)
Tetraethylene glycol monobenzyl ether

CAS Number: 86259-87-2
MDL Number: MFCD06797175
Molecular Formula: C15H24O5
Molecular Weight: 284.35
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Color: Very Pale Yellow
Specific Gravity (20/20): 1.10


Other Product Names / Synonyms: 2-[2-[2-[2-(Benzyloxy)ethoxy]ethoxy]ethoxy]ethanol

Formula: C₁₅H₂₄O₅
MW: 284.35 g/mol
Density: 1.10 (20 °C)
MDL Number: MFCD06797175
CAS Number: 86259-87-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6f509f282290d7db4fc735a5ee947a97e8ecf4d4&vid=b451d2f51f0aa505c7c272a56606841aa67c964b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Yellow to Green clear liquid
Purity (GC) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR