1,4,7,10-Tetraazacyclododecane ≥97.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: Cyclen

T1874-1G T1874-5G
TCT1874-1GEA 29 USD
TCT1874-1G TCT1874-5G
1,4,7,10-Tetraazacyclododecane ≥97.0% (by GC, titration analysis)
1,4,7,10-Tetraazacyclododecane

CAS Number: 294-90-6
Molecular Formula: C8H20N4
Molecular Weight: 172.28
Purity/Analysis Method: >97.0% (GC,T)
Form: Crystal
Color: White
Melting point (°C): 111


Product Categories: Synthetic Organic Chemistry, Ligands, Materials Chemistry, Macrocycles for Host-Guest Chemistry, Bioscience, Polyamines
Other Product Names / Synonyms: Cyclen

Formula: C₈H₂₀N₄
MW: 172.27 g/mol
Melting Pt: 111 °C
MDL Number: MFCD00066281
CAS Number: 294-90-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c0243bf52a0f7189de2c0301c74527fa529ac712&vid=c0243bf52a0f7189de2c0301c74527fa529ac712&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR