(-)-N,N,N',N'-Tetramethyl-D-tartardiamide ≥98.0% (by GC, total nitrogen)

Supplier: TCI America

T1864-5G T1864-25G
TCT1864-005GEA 130 USD
TCT1864-005G TCT1864-025G
(-)-N,N,N',N'-Tetramethyl-D-tartardiamide ≥98.0% (by GC, total nitrogen)
(-)-N,N,N',N'-Tetramethyl-D-tartardiamide

CAS Number: 63126-52-3
MDL Number: MFCD00077879
Molecular Formula: C8H16N2O4
Molecular Weight: 204.23
Purity/Analysis Method: >98.0% (GC,N)
Form: Crystal
Color: White
Melting point (°C): 188
Specific rotation [a]20/D: -46 deg (C=3, EtOH)


Other Product Names / Synonyms: (S,S)-(-)-2,3-Dihydroxy-N,N,N',N'-tetramethylsuccinamide

Formula: C₈H₁₆N₂O₄
Melting Pt: 188 °C
MDL Number: MFCD00077879
CAS Number: 63126-52-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e3078e1ae01da78b6b181a4fd84343f86e8f4b95&vid=e3078e1ae01da78b6b181a4fd84343f86e8f4b95&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR