N,N',N''-Triphenyl-1,3,5-benzenetriamine ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
Synonyms:
1,3,5-Tris(phenylamino)benzene
T1831-25G
T1831-5G
TCT1831-25GEA
224.9
USD
TCT1831-25G
TCT1831-5G
N,N',N''-Triphenyl-1,3,5-benzenetriamine ≥98.0% (by HPLC, titration analysis)
N,N',N''-Triphenyl-1,3,5-benzenetriamine
CAS Number: 102664-66-4
MDL Number: MFCD01463903
Molecular Formula: C24H21N3
Molecular Weight: 351.45
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Color: White
Melting point (°C): 198
Product Categories: Materials Chemistry, Building Blocks for Small Molecule Semiconductors
Other Product Names / Synonyms: 1,3,5-Tris(phenylamino)benzene
Formula:
C₂₄H₂₁N₃ MW: 351.45 g/mol Melting Pt: 198 °C |
MDL Number:
MFCD01463903 CAS Number: 102664-66-4 |
Specification Test Results
Purity/Analysis Method | >98.0% (HPLC,T) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...