1,4-Butandiol monovinyl ether ≥97.0% stabilized

Supplier: TCI America

Synonyms: 4-(Vinyloxy)butan-1-ol

T1796-25ML T1796-500ML
TCT1796-025MLEA 29 USD
TCT1796-025ML TCT1796-500ML
1,4-Butandiol monovinyl ether ≥97.0% stabilized
1,4-Butandiol monovinyl ether

(stabilized with KOH)
CAS Number: 17832-28-9
MDL Number: MFCD00080697
Molecular Formula: C6H12O2
Molecular Weight: 116.16
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 86
Flash Point (°C): 80
Specific Gravity (20/20): 0.95


Other Product Names / Synonyms: 4-Vinyloxybutanol

Formula: C₆H₁₂O₂
MW: 116.16 g/mol
Boiling Pt: 86 °C
Density: 0.95 (20 °C)
Flash Pt: 80 °C
MDL Number: MFCD00080697
CAS Number: 17832-28-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0696b7d13d2c08adf87959e5b62c25157186f96e&vid=0696b7d13d2c08adf87959e5b62c25157186f96e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR