1-(Trimethylsilyl)benzotriazole

Supplier: TCI America

Synonyms: 1-(Trimethylsilyl)-1H-benzotriazole

T1752-5G T1752-25G
TCT1752-005GEA 65 USD
TCT1752-005G TCT1752-25G
1-(Trimethylsilyl)benzotriazole
1-(Trimethylsilyl)benzotriazole

CAS Number: 43183-36-4
MDL Number: MFCD00216577
Molecular Formula: C9H13N3Si
Molecular Weight: 191.31
Form: Clear Liquid
Color: Colorless
Flash Point (°C): -3


Other Product Names / Synonyms: 1-TMS-1H-benzotriazole

Formula: C₉H₁₃N₃Si
Flash Pt: –3 °C
MDL Number: MFCD00216577
CAS Number: 43183-36-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=91a6536c9963ed55adeaa82ff67ca79b2ce56eb0&vid=91a6536c9963ed55adeaa82ff67ca79b2ce56eb0&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR