Tri-O-acetyl-D-galactal ≥95.0%
Supplier: TCI America
Synonyms:
(2R,3R,4R)-2-(Acetoxymethyl)-3,4-dihydro-2H-pyran-3,4-diyl diacetate, 1,5-Anhydro-2-deoxy-D-lyxo-hex-1-enitol 3,4,6-Tri-O-acetyl Ether, 3,4,6-Tri-O-acetyl-D-galactal
CAS Number: 4098-06-0
MDL Number: MFCD00064092
Molecular Formula: C12H16O7
Molecular Weight: 272.25
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Color: Colorless
Flash Point (°C): 113
Specific rotation [a]20/D: -18 deg (C=1, CHCl3)
Storage Temperature: 0-10°C
Product Categories: Bioscience, Glycals, Galactose, O-Substituted Sugars
Other Product Names / Synonyms: 1,5-Anhydro-2-deoxy-D-lyxo-hex-1-enitol 3,4,6-Tri-O-acetyl Ether
Formula:
C₁₂H₁₆O₇ Flash Pt: 113 °C |
MDL Number:
MFCD00064092 CAS Number: 4098-06-0 |
|
Specification Test Results
Purity/Analysis Method | >95.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...