5,10,15,20-Tetrakis(2,4,6-trimethylphenyl)porphyrin ≥98.0% (by HPLC)
Supplier: TCI America
Synonyms:
5,10,15,20-Tetrakis(2,4,6-trimethylphenyl)-21H,23H-porphine
T1729-100MG
T1729-1G
TCT1729-100MGEA
187
USD
TCT1729-100MG
TCT1729-1G
5,10,15,20-Tetrakis(2,4,6-trimethylphenyl)porphyrin ≥98.0% (by HPLC)
5,10,15,20-Tetrakis(2,4,6-trimethylphenyl)porphyrin
CAS Number: 56396-12-4
MDL Number: MFCD01321200
Molecular Formula: C56H54N4
Molecular Weight: 783.08
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Color: Deep Purple
Product Categories: Synthetic Organic Chemistry, Ligands, Materials Chemistry, Porphyrin & Phthalocyanine Dyes (Functional Organic Dyes), Bioscience, Porphyrins
Other Product Names / Synonyms: 5,10,15,20-Tetrakis(2,4,6-trimethylphenyl)-21H,23H-porphine
Formula:
C₅₆H₅₄N₄ MW: 783.08 g/mol |
MDL Number:
MFCD01321200 CAS Number: 56396-12-4 |
Specification Test Results
Purity/Analysis Method | >98.0% (HPLC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...