Bis(2,4-dinitrophenyl) oxalate ≥98.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
Oxalic Acid Bis(2,4-dinitrophenyl) Ester, DNPO
A5303-1G
A5303-5G
TCA5303-001GEA
101
USD
TCA5303-001G
TCA5303-005G
Bis(2,4-dinitrophenyl) oxalate ≥98.0% (by titrimetric analysis)
Bis(2,4-dinitrophenyl) oxalate
[Chemiluminescence reagent for the determination of fluorescent compounds by HPLC and FIA]
CAS Number: 16536-30-4
MDL Number: MFCD00010712
Molecular Formula: C14H6N4O12
Molecular Weight: 422.22
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Product Categories: Analytical Science, Oxalates (Chemiluminescence Reagents)
Other Product Names / Synonyms: DNPO, Oxalic Acid Bis(2,4-dinitrophenyl) Ester
Formula:
C₁₄H₆N₄O₁₂ MW: 422.22 g/mol |
MDL Number:
MFCD00010712 CAS Number: 16536-30-4 |
|
Specification Test Results
Purity/Analysis Method | >98.0% (T) |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...