Tetracaine hydrochloride ≥98.0%
Supplier: TCI America
Synonyms:
2-(Dimethylamino)ethyl 4-(Butylamino)benzoate Hydrochloride, 4-(Butylamino)benzoic Acid 2-(Dimethylamino)ethyl Ester Hydrochloride
CAS Number: 136-47-0
MDL Number: MFCD00038912
Molecular Formula: C15H24N2O2
Molecular Weight: 300.83
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Color: White
Melting point (°C): 149
Product Categories: Bioscience, Other Pharmaceutical Ingredients
Other Product Names / Synonyms: 4-(Butylamino)benzoic Acid 2-(Dimethylamino)ethyl Ester Hydrochloride, 2-(Dimethylamino)ethyl 4-(Butylamino)benzoate Hydrochloride
Formula:
C₁₅H₂₄N₂O₂ Melting Pt: 149 °C |
MDL Number:
MFCD00038912 CAS Number: 136-47-0 |
|
Specification Test Results
Purity/Analysis Method | >98.0% (HPLC,T) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...