(Triisopropylsilyl)acetylene ≥95.0%

Supplier: TCI America

Synonyms: Ethynyltriisopropylsilane

T1683-5ML T1683-25ML
TCT1683-005MLEA 77 USD
TCT1683-005ML TCT1683-25ML
(Triisopropylsilyl)acetylene ≥95.0%
(Triisopropylsilyl)acetylene

CAS Number: 89343-06-6
MDL Number: MFCD00075452
Molecular Formula: C11H22Si
Molecular Weight: 182.38
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 100
Flash Point (°C): 56
Specific Gravity (20/20): 0.82

Formula: C₁₁H₂₂Si
MW: 182.38 g/mol
Boiling Pt: 100 °C
Density: 0.82 (20 °C)
Flash Pt: 56 °C
MDL Number: MFCD00075452
CAS Number: 89343-06-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1ea1036ced4c0b6cb06c3b262e5f4e606ef1a9d4&vid=1ea1036ced4c0b6cb06c3b262e5f4e606ef1a9d4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR