2-Hydroxyethyl methyl terephthalate ≥97.0% (by GC)

Supplier: TCI America

T1679-5G T1679-1G
TCT1679-005GEA 451 USD
TCT1679-005G TCT1679-1G
2-Hydroxyethyl methyl terephthalate ≥97.0% (by GC)
2-Hydroxyethyl methyl terephthalate

CAS Number: 3645-00-9
MDL Number: MFCD01321191
Molecular Formula: C11H12O5
Molecular Weight: 224.21
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Color: White
Melting point (°C): 82


Other Product Names / Synonyms: Terephthalic Acid 2-Hydroxyethyl Methyl Ester

Formula: C₁₁H₁₂O₅
Melting Pt: 82 °C
MDL Number: MFCD01321191
CAS Number: 3645-00-9
EINECS: 222-877-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=070f68879205d5e0fff3502a04eda901b6dd54cf&vid=c6f6d6840daabfb182db1d1e5563a684a7e42af3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 97.0 %
Melting point 81.0 to 84.0 deg-C
Solubility in hot Methanol very faint turbidity


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR