Methyl L-tryptophanate hydrochloride ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: H-Trp-OMe.HCl, H-Trp-OMe·HCl, L-Tryptophan methyl ester hydrochloride

T1657-25G T1657-5G
TCT1657-025GEA 104.77 USD
TCT1657-025G TCT1657-5G
Methyl L-tryptophanate hydrochloride ≥98.0% (by HPLC, titration analysis)
Methyl L-tryptophanate hydrochloride

CAS Number: 7524-52-9
MDL Number: MFCD00066134
Molecular Formula: C12H14N2O2
Molecular Weight: 254.71
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Color: White
Melting point (°C): 220
Specific rotation [a]20/D: 19.5 deg (C=5, MeOH)


Product Categories: Bioscience, Amino Acids (C-Protected)
Other Product Names / Synonyms: Methyl L-Tryptophanate Hydrochloride, H-Trp-OMe.HCl

Formula: C₁₂H₁₄N₂O₂·HCl
MW: 254.7 g/mol
Melting Pt: 220 °C
MDL Number: MFCD00066134
CAS Number: 7524-52-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ceb4d6afbb1f523432799ca44ac05a6a4063c848&vid=ceb4d6afbb1f523432799ca44ac05a6a4063c848&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR