Tris(trimethylsilyloxy)ethylene ≥95.0%

Supplier: TCI America

Synonyms: 2,2,7,7-Tetramethyl-4-((trimethylsilyl)oxy)-3,6-dioxa-2,7-disilaoct-4-ene

T1639-25G T1639-5G
TCT1639-025GEA 315.9 USD
TCT1639-025G TCT1639-005G
Tris(trimethylsilyloxy)ethylene ≥95.0%
Tris(trimethylsilyloxy)ethylene

CAS Number: 69097-20-7 MDL Number: MFCD00011641 Molecular Formula: C11H28O3Si3 Molecular Weight: 292.60 Purity/Analysis Method: 95.0% (GC) Form: Clear Liquid Color: Colorless Flash Point (°C): 39 Specific Gravity (20/20): 0.89

Formula: C₁₁H₂₈O₃Si₃
MW: 292.60 g/mol
Density: 0.89 (20 °C)
Flash Pt: 39 °C
MDL Number: MFCD00011641
CAS Number: 69097-20-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d94742b72479a9127bab6d488a71ebace0ca6bd5&vid=d94742b72479a9127bab6d488a71ebace0ca6bd5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR